Benzopinacol 95%

Supplier: Ambeed
Danger

A679586-25G A679586-5G A679586-100G
77005-862EA 22 USD
77005-862 77005-864 77602-732
Benzopinacol 95%
Benzopinacol

Precaución: for Research Use Only

Formula: (C₆H₅)₂C(OH)C(OH)(C₆H₅)₂
MDL Number: MFCD00004448
CAS Number: 464-72-2
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR