3-Chloro-2-methylbenzamide 95%

Supplier: Ambeed
Warning

A678789-1G A678789-25G A678789-5G
77005-856EA 12 USD
77005-856 77005-858 77005-860
3-Chloro-2-methylbenzamide 95%
3-Chloro-2-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₈ClNO
MDL Number: MFCD00047882
CAS Number: 205178-79-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR