4,6-Dichloro-5-(2-methoxyphenoxy)-2,2-bipyrimidine 98%

Supplier: Ambeed
Warning

Synonyms: 4,6-Dichloro-5-(2-methoxyphenoxy)-2,2'-bipyrimidine

A678592-100G A678592-10G A678592-1G A678592-25G A678592-5G
77005-846EA 509.24 USD
77005-846 77005-848 77005-850 77005-852 77005-854
4,6-Dichloro-5-(2-methoxyphenoxy)-2,2-bipyrimidine 98%
4,6-Dichloro-5-(2-methoxyphenoxy)-2,2-bipyrimidine

Precaución: for Research Use Only

Formula: C₁₅H₁₀Cl₂N₄O₂
MW: 349.17 g/mol
MDL Number: MFCD03839838
CAS Number: 150728-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR