4-(4-Chlorophenyl)dihydro-2H-pyran-2,6(3H)-dione 96%

Supplier: Ambeed
Warning

A677803-1G A677803-250MG A677803-5G
77018-512EA 85.94 USD
77018-512 77018-514 77018-516
4-(4-Chlorophenyl)dihydro-2H-pyran-2,6(3H)-dione 96%
4-(4-Chlorophenyl)dihydro-2H-pyran-2,6(3H)-dione

Precaución: for Research Use Only

Formula: C₁₁H₉ClO₃
MW: 224.64 g/mol
MDL Number: MFCD00190250
CAS Number: 53911-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR