tert-Butyl cyclopropanecarboxylate 95%

Supplier: Ambeed
Warning

A677798-10G A677798-1G A677798-250MG A677798-25G A677798-5G
76992-572EA 25 USD
76992-572 76992-574 76992-576 76992-578 76992-580
tert-Butyl cyclopropanecarboxylate 95%
tert-Butyl cyclopropanecarboxylate

Precaución: for Research Use Only

Formula: C₈H₁₄O₂
MW: 142.196 g/mol
MDL Number: MFCD09836696
CAS Number: 87661-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR