Methyl-5-(trifluoromethyl)benzo[b]thiophene-2-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-(trifluoromethyl)benzo[b]thiophene-2-carboxylate

A677144-100MG A677144-1G A677144-250MG A677144-5G
77018-504EA 26.53 USD
77018-504 77018-506 77018-508 77018-510
Methyl-5-(trifluoromethyl)benzo[b]thiophene-2-carboxylate 95%
Methyl 5-(trifluoromethyl)benzo[b]thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₇F₃O₂S
MW: 260.2323 g/mol
MDL Number: MFCD03012932
CAS Number: 146137-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR