2-Bromo-9,9-di-n-octylfluorene 97%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-9,9-dioctyl-9H-fluorene

A676929-1G A676929-5G
77018-500EA 28.65 USD
77018-500 77018-502
2-Bromo-9,9-di-n-octylfluorene 97%
2-Bromo-9,9-di-n-octylfluorene

Precaución: for Research Use Only

Formula: C₂₉H₄₁Br
MW: 469.55 g/mol
MDL Number: MFCD12024273
CAS Number: 302554-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR