2-(Trifluoromethyl)-1H-imidazole 97%

Supplier: Ambeed
Warning

A676729-100MG A676729-1G A676729-250MG A676729-5G
77035-646EA 14 USD
77035-646 77035-648 77035-650 77035-652
2-(Trifluoromethyl)-1H-imidazole 97%
2-(Trifluoromethyl)-1H-imidazole

Precaución: for Research Use Only

Formula: C₄H₃F₃N₂
MDL Number: MFCD18415320
CAS Number: 66675-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR