4-(1,3-Oxazol-5-yl)phenol 97%

Supplier: Ambeed
Warning

Synonyms: 4-(5-Oxazolyl)phenol

A676621-100MG A676621-1G
77005-842EA 13.8 USD
77005-842 77005-844
4-(1,3-Oxazol-5-yl)phenol 97%
4-(1,3-Oxazol-5-yl)phenol

Precaución: for Research Use Only

Formula: C₉H₇NO₂
MW: 161.157 g/mol
MDL Number: MFCD00207683
CAS Number: 1128-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR