(S,S)-(+)-2-Amino-1-phenylpropyldiphenylphosphine ≥97%

Supplier: Ambeed
Warning

Synonyms: (1S,2S)-1-(Diphenylphosphino)-1-phenylpropan-2-amine, (1S,2S)-(2-Diphenylphosphino)-1-methyl-2-phenylethylamine, (1S,2S)-2-Amino-1-phenylpropyldiphenylphosphine

A676074-100MG A676074-250MG
76992-568EA 68.96 USD
76992-568 76992-570
(S,S)-(+)-2-Amino-1-phenylpropyldiphenylphosphine ≥97%
(S,S)-(+)-2-Amino-1-phenylpropyldiphenylphosphine

Precaución: for Research Use Only

Formula: C₂₁H₂₂NP
MW: 319.388 g/mol
MDL Number: MFCD11044864
CAS Number: 341968-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR