Bis[rhodium(alpha,alpha,alpha',alpha'-tetramethyl-1,3-benzenedipropionic acid)] 98%

Supplier: Ambeed
Warning

A676065-100MG A676065-1G A676065-250MG A676065-50MG A676065-5G
76992-558EA 44 USD
76992-558 76992-560 76992-562 76992-564 76992-566
Bis[rhodium(alpha,alpha,alpha',alpha'-tetramethyl-1,3-benzenedipropionic acid)] 98%
Bis[rhodium(alpha,alpha,alpha',alpha'-tetramethyl-1,3-benzenedipropionic acid)]

Precaución: for Research Use Only

Formula: C₃₂H₄₀O₈Rh₂
MDL Number: MFCD08457636
CAS Number: 819050-89-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR