Ethyl-4-bromo-1,5-dimethyl-1H-pyrazole-3-carboxylate 97%

Supplier: Ambeed
Danger

Synonyms: Ethyl 4-Bromo-1,5-dimethyl-1H-pyrazole-3-carboxylate

A675751-100MG A675751-10G A675751-1G A675751-250MG A675751-5G
77005-832EA 33.95 USD
77005-832 77005-834 77005-836 77005-838 77005-840
Ethyl-4-bromo-1,5-dimethyl-1H-pyrazole-3-carboxylate 97%
Ethyl 4-bromo-1,5-dimethyl-1H-pyrazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₁BrN₂O₂
MW: 247.0891 g/mol
MDL Number: MFCD00207970
CAS Number: 5775-90-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR