Boc-isoguvacine 98%

Supplier: Ambeed
Danger

Synonyms: 1-Boc-1,2,3,6-tetrahydropyridine-4-carboxylic Acid

A675533-100MG A675533-1G A675533-250MG A675533-5G
77018-486EA 15.92 USD
77018-486 77018-488 77018-490 77614-086
Boc-isoguvacine 98%
Boc-isoguvacine

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO₄
MW: 227.26 g/mol
MDL Number: MFCD02682417
CAS Number: 70684-84-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR