6-Methoxy-5-nitro (1H)indazole 95%

Supplier: Ambeed
Warning

Synonyms: 6-Methoxy-5-nitro-1H-indazole

A674698-1G A674698-250MG A674698-5G
76992-544EA 234.46 USD
76992-544 76992-546 77614-082
6-Methoxy-5-nitro (1H)indazole 95%
6-Methoxy-5-nitro (1H)indazole

Precaución: for Research Use Only

Formula: C₈H₇N₃O₃
MW: 193.159 g/mol
MDL Number: MFCD11007881
CAS Number: 152626-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR