2-Chloro-3-nitroisonicotinic acid 97%

Supplier: Ambeed
Warning

A673812-1G A673812-250MG
77018-482EA 29.71 USD
77018-482 77018-484
2-Chloro-3-nitroisonicotinic acid 97%
2-Chloro-3-nitroisonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₃ClN₂O₄
MW: 202.55 g/mol
MDL Number: MFCD01928336
CAS Number: 353281-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR