5-Methylquinoline 98%

Supplier: Ambeed
Warning

A673640-10G A673640-1G A673640-25G A673640-5G
77035-636EA 95.49 USD
77035-636 77035-638 77035-640 77035-642
5-Methylquinoline 98%
5-Methylquinoline

Precaución: for Research Use Only

Formula: C₁₀H₉N
MW: 143.19 g/mol
MDL Number: MFCD01685457
CAS Number: 7661-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR