Hexadecan-3-ol 98%

Supplier: Ambeed
Warning

A672714-100MG A672714-1G A672714-250MG
77035-626EA 26 USD
77035-626 77035-628 77035-630
Hexadecan-3-ol 98%
Hexadecan-3-ol

Precaución: for Research Use Only

Formula: C₁₆H₃₄O
MDL Number: MFCD00051480
CAS Number: 593-03-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR