2,3-O-Isopropylidene-α,β-D-ribofuranose 95% (by GC)

Supplier: Ambeed
Warning

Synonyms: (4R,5R)-5-((R)-1,2-Dihydroxyethyl)-2,2-dimethyl-1,3-dioxolane-4-carbaldehyde

A672427-100MG A672427-1G A672427-250MG
76992-526EA 13.8 USD
76992-526 76992-528 76992-530
2,3-O-Isopropylidene-α,β-D-ribofuranose 95% (by GC)
2,3-O-Isopropylidene-alpha,beta-D-ribofuranose

Precaución: for Research Use Only

Formula: C₈H₁₄O₅
MW: 190.20 g/mol
MDL Number: MFCD01075718
CAS Number: 13199-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR