4,4'-Bipiperidine 98%

Supplier: Ambeed
Warning

A672305-100MG A672305-1G A672305-250MG A672305-5G
77035-618EA 31 USD
77035-618 77035-620 77035-622 77873-074
4,4'-Bipiperidine 98%
4,4'-Bipiperidine

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂
MDL Number: MFCD01104315
CAS Number: 15336-72-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR