Methyl-4-bromo-1-methyl-1H-pyrazole-5-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-bromo-1-methyl-1H-pyrazole-5-carboxylate

A670943-1G A670943-250MG A670943-25G A670943-5G
77035-602EA 43.5 USD
77035-602 77035-604 77035-606 77035-608
Methyl-4-bromo-1-methyl-1H-pyrazole-5-carboxylate 98%
Methyl-4-bromo-1-methyl-1H-pyrazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₆H₇BrN₂O₂
MW: 219.04 g/mol
MDL Number: MFCD01459850
CAS Number: 514816-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR