O-2-Propynylhydroxylamine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: O-(Prop-2-yn-1-yl)hydroxylamine hydrochloride

A670545-100MG A670545-250MG A670545-1G
76992-522EA 64.72 USD
76992-522 76992-524 77684-576
O-2-Propynylhydroxylamine hydrochloride 98%
O-2-Propynylhydroxylamine hydrochloride

Precaución: For research use only.

Formula: C₃H₆ClNO
MW: 107.54 g/mol
MDL Number: MFCD12912207
CAS Number: 21663-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR