(4-Biphenylylmethyl)hydrazine hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: ([1,1'-Biphenyl]-4-ylmethyl)hydrazine hydrochloride

A670525-100MG A670525-1G A670525-250MG
76992-516EA 16 USD
76992-516 76992-518 76992-520
(4-Biphenylylmethyl)hydrazine hydrochloride 95%
(4-Biphenylylmethyl)hydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₁₄N₂·ClH
MDL Number: MFCD01722362
CAS Number: 75333-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR