tert-Butyl (6-fluoropyridin-3-yl)carbamate ≥97%

Supplier: Ambeed
Warning

A670443-100MG A670443-1G A670443-250MG
77018-464EA 9 USD
77018-464 77018-466 77891-326
tert-Butyl (6-fluoropyridin-3-yl)carbamate ≥97%
tert-Butyl (6-fluoropyridin-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₃FN₂O₂
MDL Number: MFCD08274661
CAS Number: 171178-41-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR