(4-(Allylcarbamoyl)phenyl)boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Allylaminocarbonylphenylboronic acid

A669706-1G A669706-250MG A669706-5G
77018-458EA 98 USD
77018-458 77018-460 77018-462
(4-(Allylcarbamoyl)phenyl)boronic acid 97%
(4-(Allylcarbamoyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂BNO₃
MDL Number: MFCD04115674
CAS Number: 850568-20-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR