(E)-3-(m-Tolyl)acrylaldehyde 95%
Supplier: Ambeed
|
Warning
|
A669442-100MG
A669442-1G
A669442-250MG
A669442-5G
76992-504EA
8
USD
76992-504
76992-506
76992-508
76992-510
(E)-3-(m-Tolyl)acrylaldehyde 95%
(E)-3-(m-Tolyl)acrylaldehyde
Precaución: for Research Use Only
Formula:
C₁₀H₁₀O MW: 146.18 g/mol |
MDL Number:
MFCD08460293 CAS Number: 93614-80-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...