Boc-Lys(fmoc)-OMe 98%

Supplier: Ambeed
Warning

Synonyms: (S)-Methyl 6-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-2-((tert-butoxycarbonyl)amino)hexanoate, Nα-Boc-Nepsilon-Fmoc-L-lysine methyl ester

A669181-1G A669181-250MG
76992-500EA 108.22 USD
76992-500 76992-502
Boc-Lys(fmoc)-OMe 98%
Boc-Lys(fmoc)-OMe

Precaución: for Research Use Only

Formula: C₂₇H₃₄N₂O₆
MW: 482.577 g/mol
MDL Number: MFCD00236856
CAS Number: 133628-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR