6-Methyl-1H-benzo[d]imidazol-7-amine 95+%
Supplier: Ambeed
|
Danger
|
A668609-100MG
A668609-250MG
77018-452EA
219.61
USD
77018-452
77018-454
6-Methyl-1H-benzo[d]imidazol-7-amine 95+%
6-Methyl-1H-benzo[d]imidazol-7-amine
Precaución: for Research Use Only
Formula:
C₈H₉N₃ MW: 147.17 g/mol |
MDL Number:
MFCD09754321 CAS Number: 935873-39-5 UN: 3259 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...