Methyl 6-cyanonicotinate 97%

Supplier: Ambeed
Warning

A668004-1G A668004-5G
77005-778EA 15.92 USD
77005-778 77005-780 77005-782 77005-784
Methyl 6-cyanonicotinate 97%
Methyl 6-cyanonicotinate

Precaución: for Research Use Only

Formula: C₈H₆N₂O₂
MDL Number: MFCD09927471
CAS Number: 89809-65-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR