Ethyl 5-methoxy-1H-indazole-3-carboxylate 98%

Supplier: Ambeed
Warning

A667178-100MG A667178-1G A667178-250MG
77018-432EA 41.38 USD
77018-432 77018-434 77018-436
Ethyl 5-methoxy-1H-indazole-3-carboxylate 98%
Ethyl 5-methoxy-1H-indazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂O₃
MW: 220.22 g/mol
MDL Number: MFCD06659782
CAS Number: 865887-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR