4-Bromobenzyl methyl ether 97%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-4-(methoxymethyl)benzene

A666808-100G A666808-1G A666808-5G
76992-496EA 883.73 USD
76992-496 76992-498 77614-056
4-Bromobenzyl methyl ether 97%
4-Bromobenzyl methyl ether

Precaución: for Research Use Only

Formula: C₈H₉BrO
MW: 201.0600 g/mol
MDL Number: MFCD09265134
CAS Number: 1515-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR