4-Iodo-2,6-dimethylaniline hydrochloride 97%

Supplier: Ambeed
Warning

A666319-1G A666319-250MG
76992-492EA 33 USD
76992-492 76992-494
4-Iodo-2,6-dimethylaniline hydrochloride 97%
4-Iodo-2,6-dimethylaniline hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₁ClIN
MDL Number: MFCD19442006
CAS Number: 138385-59-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR