1-(4-Nitrophenyl)cyclopropanecarbonitrile 98%

Supplier: Ambeed
Warning

A665951-100MG A665951-1G A665951-250MG
76992-486EA 27 USD
76992-486 76992-488 76992-490
1-(4-Nitrophenyl)cyclopropanecarbonitrile 98%
1-(4-Nitrophenyl)cyclopropanecarbonitrile

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₂
MDL Number: MFCD07374412
CAS Number: 408328-42-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR