Dicyclohexylamine (S)-6-(((benzyloxy)carbonyl)amino)-2-((tert-butoxycarbonyl)amino)hexanoate 97%

Supplier: Ambeed
Warning

A665828-100G A665828-25G A665828-5G
77005-764EA 170 USD
77005-764 77005-766 77005-768
Dicyclohexylamine (S)-6-(((benzyloxy)carbonyl)amino)-2-((tert-butoxycarbonyl)amino)hexanoate 97%
Dicyclohexylamine (S)-6-(((benzyloxy)carbonyl)amino)-2-((tert-butoxycarbonyl)amino)hexanoate

Precaución: for Research Use Only

Formula: C₃₁H₅₁N₃O₆
MDL Number: MFCD00066511
CAS Number: 16948-04-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR