2-Bromo-6-(tert-butoxy)pyridine 97%

Supplier: Ambeed
Warning

A665687-1G A665687-5G
76992-482EA 49.87 USD
76992-482 76992-484
2-Bromo-6-(tert-butoxy)pyridine 97%
2-Bromo-6-(tert-butoxy)pyridine

Precaución: for Research Use Only

Formula: C₉H₁₂BrNO
MW: 230.10 g/mol
MDL Number: MFCD00834991
CAS Number: 949160-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR