BIX2188 Z-Isomer 98%

Supplier: Ambeed
Warning

Synonyms: BIX-02188, BIX02188, (Z)-3-(((3-((dimethylamino)methyl)phenyl)amino)(phenyl)methylene)-2-oxoindoline-6-carboxamide

A665681-10MG A665681-1MG A665681-5MG
77035-558EA 62 USD
77035-558 77035-560 77035-562
BIX2188 Z-Isomer 98%
BIX2188 Z-Isomer

Precaución: for Research Use Only

Formula: C₂₅H₂₄N₄O₂
MDL Number: MFCD18157583
CAS Number: 334949-59-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR