4-Amino-2-chlorobenzamide 98%

Supplier: Ambeed
Warning

A665645-1G A665645-25G A665645-5G
77018-424EA 35 USD
77018-424 77018-426 77018-428
4-Amino-2-chlorobenzamide 98%
4-Amino-2-chlorobenzamide

Precaución: for Research Use Only

Formula: C₇H₇ClN₂O
MDL Number: MFCD09043062
CAS Number: 211374-81-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR