4-(4-Bromobenzoyl)piperidine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: (4-Bromophenyl)(4-piperidyl)methanone Hydrochloride

A665148-5G A665148-1G
76992-480EA 254.62 USD
76992-480 77684-565
4-(4-Bromobenzoyl)piperidine hydrochloride 97%
4-(4-Bromobenzoyl)piperidine hydrochloride

Precaución: For research use only.

Formula: C₁₂H₁₅BrClNO
MW: 304.6100 g/mol
MDL Number: MFCD06804519
CAS Number: 64671-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR