4-Nitro-2-(trifluoromethyl)benzaldehyde 95%

Supplier: Ambeed
Warning

A664453-5G A664453-250MG
76992-478EA 225 USD
76992-478 77891-315
4-Nitro-2-(trifluoromethyl)benzaldehyde 95%
4-Nitro-2-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄F₃NO₃
MDL Number: MFCD13185769
CAS Number: 50551-17-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR