Pentaerythrityl tetrachloride 98%

Supplier: Ambeed
Warning

Synonyms: Pentaerythrityl Tetrachloride

A664360-25G A664360-1G
77018-422EA 124 USD
77018-422 77893-342
Pentaerythrityl tetrachloride 98%
Pentaerythrityl tetrachloride

Precaución: for Research Use Only

Formula: C₅H₈Cl₄
MW: 209.93 g/mol
Boiling Pt: 233
Melting Pt: 95
MDL Number: MFCD00000942
CAS Number: 3228-99-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR