3-Bromoindolin-2-one 98%

Supplier: Ambeed
Warning

A664285-100MG A664285-1G
77018-418EA 15 USD
77018-418 77018-420
3-Bromoindolin-2-one 98%
3-Bromoindolin-2-one

Precaución: for Research Use Only

Formula: C₈H₆BrNO
MDL Number: MFCD00474900
CAS Number: 22942-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR