Methyl 4-(trichloromethyl)benzoate 95%

Supplier: Ambeed
Warning

A664244-1G A664244-25G A664244-5G
76992-472EA 13 USD
76992-472 76992-474 76992-476
Methyl 4-(trichloromethyl)benzoate 95%
Methyl 4-(trichloromethyl)benzoate

Precaución: for Research Use Only

Formula: C₉H₇Cl₃O₂
MDL Number: MFCD00511226
CAS Number: 14815-87-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR