(R)-4-(2-(2-(2-methylpyrrolidin-1-yl)ethyl)benzofuran-5-yl)benzonitrile 95%

Supplier: Ambeed
Warning

A663348-10MG A663348-1MG A663348-25MG A663348-50MG A663348-5MG
77035-538EA 237.65 USD
77035-538 77035-540 77035-542 77035-544 77035-546
(R)-4-(2-(2-(2-methylpyrrolidin-1-yl)ethyl)benzofuran-5-yl)benzonitrile 95%
(R)-4-(2-(2-(2-methylpyrrolidin-1-yl)ethyl)benzofuran-5-yl)benzonitrile

Precaución: for Research Use Only

Formula: C₂₂H₂₂N₂O
MDL Number: MFCD13248643
CAS Number: 460746-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR