Ethyl 7-chloroquinoline-2-carboxylate 95+%

Supplier: Ambeed
Warning

A663163-100MG A663163-250MG A663163-1G
77018-414EA 14 USD
77018-414 77018-416 77891-312
Ethyl 7-chloroquinoline-2-carboxylate 95+%
Ethyl 7-chloroquinoline-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₀ClNO₂
MDL Number: MFCD27952681
CAS Number: 116529-90-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR