tert-Butyl 7-hydroxy-2-azaspiro[3.5]nonane-2-carboxylate 95%

Supplier: Ambeed
Warning

A662965-1G A662965-100MG A662965-250MG
77018-412EA 334.19 USD
77018-412 77684-559 77684-560
tert-Butyl 7-hydroxy-2-azaspiro[3.5]nonane-2-carboxylate 95%
tert-Butyl 7-hydroxy-2-azaspiro[3.5]nonane-2-carboxylate

Precaución: For research use only.

Formula: C₁₃H₂₃NO₃
MW: 241.32 g/mol
MDL Number: MFCD20926146
CAS Number: 1363383-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR