5-Isopropyl-4-nitro-1H-pyrazole-3-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
A662856-1G
A662856-250MG
77035-534EA
310.85
USD
77035-534
77035-536
5-Isopropyl-4-nitro-1H-pyrazole-3-carboxylic acid 97%
5-Isopropyl-4-nitro-1H-pyrazole-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C₇H₉N₃O₄ MW: 199.16 g/mol |
MDL Number:
MFCD09052668 CAS Number: 141721-97-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...