1,4-Diazaspiro[4.5]decan-2-one 95%

Supplier: Ambeed
Warning

A661361-100MG A661361-1G A661361-250MG
77005-758EA 61 USD
77005-758 77005-760 77005-762
1,4-Diazaspiro[4.5]decan-2-one 95%
1,4-Diazaspiro[4.5]decan-2-one

Precaución: for Research Use Only

Formula: C₈H₁₄N₂O
MDL Number: MFCD14584836
CAS Number: 19718-88-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR