2-(3'-Chlorobenzyloxy)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (2-((3-Chlorobenzyl)oxy)phenyl)boronic acid

A661309-100MG A661309-1G A661309-250MG
77018-400EA 33 USD
77018-400 77018-402 77018-404
2-(3'-Chlorobenzyloxy)phenylboronic acid 98%
2-(3'-Chlorobenzyloxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₂BClO₃
MDL Number: MFCD06411351
CAS Number: 849062-32-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR