1-(3,4-Dimethoxy-5-nitrophenyl)ethanone 95%

Supplier: Ambeed
Warning

A661275-1G A661275-250MG
77018-394EA 30.77 USD
77018-394 77018-396 77018-398
1-(3,4-Dimethoxy-5-nitrophenyl)ethanone 95%
1-(3,4-Dimethoxy-5-nitrophenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₅
MDL Number: MFCD03932014
CAS Number: 134610-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR