2-Methylpyrido[3,4-d]pyrimidin-4(1H)-one 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A661197-1G A661197-250MG
76992-462EA 169.75 USD
76992-462 77684-556
2-Methylpyrido[3,4-d]pyrimidin-4(1H)-one 98%, Ambeed.Inc
2-Methylpyrido[3,4-d]pyrimidin-4(1H)-one

Precaución: For research use only.

Formula: C₈H₇N₃O
MW: 161.16 g/mol
MDL Number: MFCD26793523
CAS Number: 22389-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR