2,3,4,5-Tetrafluorophenylboronic acid 95+%

Supplier: Ambeed
Warning

Synonyms: 2,3,4,5-Tetrafluorobenzeneboronic acid, Dihydroxy(2,3,4,5-tetrafluorophenyl)borane, (2,3,4,5-Tetrafluorophenyl)metaboric acid, (2,3,4,5-Tetrafluorophenyl)boronic acid

A661037-1G A661037-250MG A661037-5G
77005-752EA 207.94 USD
77005-752 77005-754 77005-756
2,3,4,5-Tetrafluorophenylboronic acid 95+%
2,3,4,5-Tetrafluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₃BF₄O₂
MW: 193.89 g/mol
MDL Number: MFCD04039308
CAS Number: 179923-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR